p-Tolunitrile, 98+%, Thermo Scientific Chemicals
p-Tolunitrile, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Tolunitrile, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 158022500
Price (USD)/ Each
112.00
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
112.00
Add to cart
p-Tolunitrile, 98+%, Thermo Scientific Chemicals
Catalog number158022500
Price (USD)/ Each
112.00
-
Add to cart
Chemical Identifiers
CAS104-85-8
IUPAC Name4-methylbenzonitrile
Molecular FormulaC8H7N
InChI KeyVCZNNAKNUVJVGX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid or solid
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only