1,2-Diaminocyclohexane, 99%, mixture of cis and trans, Thermo Scientific Chemicals
1,2-Diaminocyclohexane, 99%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Diaminocyclohexane, 99%, mixture of cis and trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 158100010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS694-83-7
IUPAC Name(1R,2S)-cyclohexane-1,2-bis(aminium)
Molecular FormulaC6H16N2
InChI KeySSJXIUAHEKJCMH-OLQVQODUSA-P
SMILES[NH3+][C@H]1CCCC[C@H]1[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<75 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4860 to 1.4920 (20°C, 589 nm)
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only