Thermo Scientific Chemicals

1,2-Diaminocyclohexane, 99%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 158102500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Diaminocyclohexane, 99%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 158102500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 158102500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
694-83-7
IUPAC Name
(1R,2S)-cyclohexane-1,2-bis(aminium)
Molecular Formula
C6H16N2
InChI Key
SSJXIUAHEKJCMH-OLQVQODUSA-P
SMILES
[NH3+][C@H]1CCCC[C@H]1[NH3+]
Color scale
=<75 APHA
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4860 to 1.4920 (20°C, 589 nm)
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text