trans-2-Hexenal, 99%, Thermo Scientific Chemicals
trans-2-Hexenal, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-Hexenal, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number 158131000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6728-26-3
IUPAC Name(2E)-hex-2-enal
Molecular FormulaC6H10O
InChI KeyMBDOYVRWFFCFHM-SNAWJCMRSA-N
SMILESCCC\C=C\C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4440 to 1.4500 (20°C, 589 nm)
Density(20°C) 0.843 to 0.849 g/ml
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only