Thermo Scientific Chemicals

4,4'-Isopropylidenediphenol, 97%, Thermo Scientific Chemicals

Catalog number: 158245000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

4,4'-Isopropylidenediphenol, 97%, Thermo Scientific Chemicals

Catalog number: 158245000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 158245000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
80-05-7
IUPAC Name
4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
Molecular Formula
C15H16O2
InChI Key
IISBACLAFKSPIT-UHFFFAOYSA-N
SMILES
CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Appearance (Color)
White
Appearance (Form)
Flakes or prills
GC
>=96.0 %
Infrared spectrum
Conforms
Melting point
155°C to 160°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text