n-Propyl acetate, 99%, pure, Thermo Scientific Chemicals
n-Propyl acetate, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Propyl acetate, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 158290025
Price (USD)/ Each
169.65
Online exclusive
188.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
169.65
Online exclusive
188.00 
Save 18.35 (10%)
Add to cart
n-Propyl acetate, 99%, pure, Thermo Scientific Chemicals
Catalog number158290025
Price (USD)/ Each
169.65
Online exclusive
188.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS109-60-4
IUPAC Namepropyl acetate
Molecular FormulaC5H10O2
InChI KeyYKYONYBAUNKHLG-UHFFFAOYSA-N
SMILESCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
Refractive index1.3830 to 1.3850 (20°C, 589 nm)
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only