Thermo Scientific Chemicals

n-Propyl acetate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 158290100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

n-Propyl acetate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 158290100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 158290100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-60-4
IUPAC Name
propyl acetate
Molecular Formula
C5H10O2
InChI Key
YKYONYBAUNKHLG-UHFFFAOYSA-N
SMILES
CCCOC(C)=O
Appearance (Form)
Clear liquid
Color scale
=<15 APHA
Infrared spectrum
Conforms
Refractive index
1.3830 to 1.3850 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text