Pyridinium bromide perbromide, 90+%, technical
Pyridinium bromide perbromide, 90+%, technical
Pyridinium bromide perbromide, 90+%, technical
Pyridinium bromide perbromide, 90+%, technical
Thermo Scientific Chemicals

Pyridinium bromide perbromide, 90+%, technical

CAS: 39416-48-3 | C5H6Br3N | 319.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 158365000
Price (USD)
293.65
Special offer
Online exclusive
Ends: 26-Sep-2025
362.00
Save 68.35 (19%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
293.65
Special offer
Online exclusive
Ends: 26-Sep-2025
362.00
Save 68.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS39416-48-3
Molecular FormulaC5H6Br3N
InChI KeyGWJBDYRGIFGTOG-UHFFFAOYSA-O
SMILES[H+].[Br].[Br].[Br].C1=CC=NC=C1
Molecular Weight (g/mol)319.82
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Infrared spectrumConforms
Melting point130°C to 137°C (Dec)
Appearance (Form)Powder or crystals and/or chunks
Titration Iodometric>=90.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link