1x1 image pixel for data collection
Benzyl isocyanide, 98%
Benzyl isocyanide, 98%
Benzyl isocyanide, 98%
Benzyl isocyanide, 98%
Thermo Scientific Chemicals

Benzyl isocyanide, 98%

CAS: 10340-91-7 | C8H7N | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 158390050
Price (USD)
271.65
Special offer
Online exclusive
319.00
Save 47.35 (15%)
Each
In stock
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
271.65
Special offer
Online exclusive
319.00
Save 47.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS10340-91-7
IUPAC Name(isocyanomethyl)benzene
Molecular FormulaC8H7N
InChI KeyRIWNFZUWWRVGEU-UHFFFAOYSA-N
SMILES[C-]#[N+]CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Refractive index1.5200 to 1.5220 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless to dark yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection