Cyclohexyl isocyanide, 99%, Thermo Scientific Chemicals
Cyclohexyl isocyanide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexyl isocyanide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 158400010
Price (USD)/ Each
43.65
Online exclusive
47.90 
Save 4.25 (9%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
47.90 
Save 4.25 (9%)
Add to cart
Cyclohexyl isocyanide, 99%, Thermo Scientific Chemicals
Catalog number158400010
Price (USD)/ Each
43.65
Online exclusive
47.90 
Save 4.25 (9%)
-
Add to cart
Chemical Identifiers
IUPAC Nameisocyanocyclohexane
Molecular FormulaC7H11N
InChI KeyXYZMOVWWVXBHDP-UHFFFAOYSA-N
SMILES[C-]#[N+]C1CCCCC1
Molecular Weight (g/mol)109.17
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to light brown
Refractive index1.4490 to 1.4520 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only