Formanilide, 99+%, Thermo Scientific Chemicals
Formanilide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Formanilide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 158425000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS103-70-8
IUPAC NameN-phenylformamide
Molecular FormulaC7H7NO
InChI KeyDYDNPESBYVVLBO-UHFFFAOYSA-N
SMILESO=CNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey-beige
Appearance (Form)Crystals
Melting point46°C to 49°C
GC>=99.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only