Thermo Scientific Chemicals

4-Acetamidobenzoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 158430250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Acetamidobenzoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 158430250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 158430250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
556-08-1
IUPAC Name
4-acetamidobenzoic acid
Molecular Formula
C9H9NO3
InChI Key
QCXJEYYXVJIFCE-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=C(C=C1)C(O)=O
HPLC
>=99.0 %
Appearance (Color)
Off-white to light beige
Infrared spectrum
Conforms
Titration with NaOH
99.0 to 101.0 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text