4-Acetamidobenzoic acid, 99+%, Thermo Scientific Chemicals
4-Acetamidobenzoic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetamidobenzoic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 158431000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS556-08-1
IUPAC Name4-acetamidobenzoic acid
Molecular FormulaC9H9NO3
InChI KeyQCXJEYYXVJIFCE-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Appearance (Color)Off-white to light beige
Infrared spectrumConforms
Titration with NaOH99.0 to 101.0 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only