Thermo Scientific Chemicals

1,12-Dibromododecane, 96%, Thermo Scientific Chemicals

Catalog number: 158451000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,12-Dibromododecane, 96%, Thermo Scientific Chemicals

Catalog number: 158451000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 158451000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3344-70-5
IUPAC Name
1,12-dibromododecane
Molecular Formula
C12H24Br2
InChI Key
ZJJATABWMGVVRZ-UHFFFAOYSA-N
SMILES
BrCCCCCCCCCCCCBr
Appearance (Form)
Crystalline solid
Melting point
37°C to 42°C
Appearance (Color)
Off-white to light brown
GC
>=95 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text