Thermo Scientific Chemicals

1,2-Dibromotetrachloroethane, 97%, Thermo Scientific Chemicals

Catalog number: 158460250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Dibromotetrachloroethane, 97%, Thermo Scientific Chemicals

Catalog number: 158460250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 158460250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
630-25-1
IUPAC Name
1,2-dibromo-1,1,2,2-tetrachloroethane
Molecular Formula
C2Br2Cl4
InChI Key
WJUKOGPNGRUXMG-UHFFFAOYSA-N
SMILES
ClC(Cl)(Br)C(Cl)(Cl)Br
Appearance (Color)
White to beige
GC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text