Thermo Scientific Chemicals

4-Amino-1-naphthol hydrochloride, 90%, technical, Thermo Scientific Chemicals

Catalog number: 158510050
5 g, Each
Thermo Scientific Chemicals

4-Amino-1-naphthol hydrochloride, 90%, technical, Thermo Scientific Chemicals

Catalog number: 158510050
5 g, Each
Quantity
Catalog number: 158510050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5959-56-8
IUPAC Name
4-hydroxynaphthalen-1-aminium chloride
Molecular Formula
C10H10ClNO
InChI Key
FDBQTRARWCKEJY-UHFFFAOYSA-N
SMILES
[Cl-].[NH3+]C1=C2C=CC=CC2=C(O)C=C1
Appearance (Color)
Pink to grey
Infrared spectrum
Conforms
Appearance (Form)
Powder
HPLC
>=88 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text