2-Amino-5-nitrothiazole, 97%, Thermo Scientific Chemicals
2-Amino-5-nitrothiazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-nitrothiazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 158521000
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.10 
Save 13.45 (14%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.10 
Save 13.45 (14%)
Add to cart
2-Amino-5-nitrothiazole, 97%, Thermo Scientific Chemicals
Catalog number158521000
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.10 
Save 13.45 (14%)
-
Add to cart
Chemical Identifiers
CAS121-66-4
IUPAC Name5-nitro-1,3-thiazol-2-amine
Molecular FormulaC3H3N3O2S
InChI KeyMIHADVKEHAFNPG-UHFFFAOYSA-N
SMILESNC1=NC=C(S1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=96.0 %
Appearance (Color)Yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only