Thermo Scientific Chemicals

4-Chlorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 158560250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Chlorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 158560250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 158560250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
98-60-2
IUPAC Name
4-chlorobenzene-1-sulfonyl chloride
Molecular Formula
C6H4Cl2O2S
InChI Key
ZLYBFBAHAQEEQQ-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)S(Cl)(=O)=O
Appearance (Form)
Powder or crystals and/or chunks
Melting point
50°C to 55°C
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow
Titration Argentometric
96.0 to 104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text