trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
100 g
500 g
Packaging:
View all
Plastic drum
Catalog number 158570025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS140-10-3
IUPAC Name(2E)-3-phenylprop-2-enoic acid
Molecular FormulaC9H8O2
InChI KeyWBYWAXJHAXSJNI-VOTSOKGWSA-N
SMILESOC(=O)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Appearance (Color)White to light yellow
Melting point131°C to 136°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only