1,2-Propanediol, 99%, extra pure
1,2-Propanediol, 99%, extra pure
1,2-Propanediol, 99%, extra pure
1,2-Propanediol, 99%, extra pure
Thermo Scientific Chemicals

1,2-Propanediol, 99%, extra pure

CAS: 57-55-6 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 158720010
Price (USD)
56.65
Online Exclusive
62.60
Save 5.95 (10%)
Each
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.60
Save 5.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS57-55-6
IUPAC Namepropane-1,2-diol
Molecular FormulaC3H8O2
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILESCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4310 to 1.4330 (20°C, 589 nm)
Appearance (Form)Clear viscous liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link