Thermo Scientific Chemicals

1,2-Propanediol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 158720250
25 L, Metal drum, Each
Thermo Scientific Chemicals

1,2-Propanediol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 158720250
25 L, Metal drum, Each
Quantity
Packaging
Catalog number: 158720250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
57-55-6
IUPAC Name
propane-1,2-diol
Molecular Formula
C3H8O2
InChI Key
DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES
CC(O)CO
Water
=<0.2 % (Coulometric)
Refractive index
1.4310 to 1.4330 (20°C, 589 nm)
Appearance (Form)
Clear viscous liquid
Color scale
=<10 APHA
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text