Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Succinic Acid 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 158740010
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
Add to cart
Succinic Acid 99%, Thermo Scientific Chemicals
Catalog number158740010
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS110-15-6
IUPAC Namebutanedioic acid
Molecular FormulaC4H6O4
InChI KeyKDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILESOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Fumaric and maleic acid=<1 %
Appearance (Form)Crystalline powder
Melting point185°C to 190°C
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only