Thermo Scientific Chemicals

Succinic Acid 99%, Thermo Scientific Chemicals

Catalog number: 158742500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Succinic Acid 99%, Thermo Scientific Chemicals

Catalog number: 158742500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 158742500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-15-6
IUPAC Name
butanedioic acid
Molecular Formula
C4H6O4
InChI Key
KDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILES
OC(=O)CCC(O)=O
Titration with NaOH
>=98.5 %
Fumaric and maleic acid
=<1 %
Appearance (Form)
Crystalline powder
Melting point
185°C to 190°C
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text