Thermo Scientific Chemicals

Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals

Catalog number: 158751000
100 g, Each
Thermo Scientific Chemicals

Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals

Catalog number: 158751000
100 g, Each
Quantity
Catalog number: 158751000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6106-21-4
IUPAC Name
disodium butanedioate hexahydrate
Molecular Formula
C4H16Na2O10
InChI Key
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O
Appearance (Color)
White
Solubility
(10 % in water) Clear
Appearance (Form)
Crystalline powder or crystals or granules
Titration with HClO4
96.0 to 103.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text