Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals
Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Catalog number 158755000
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.70 
Save 13.05 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.70 
Save 13.05 (15%)
Add to cart
Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals
Catalog number158755000
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.70 
Save 13.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium butanedioate hexahydrate
Molecular FormulaC4H16Na2O10
InChI KeyZBTUYCUNQBRXOR-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O
Molecular Weight (g/mol)270.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Solubility(10 % in water) Clear
Appearance (Form)Crystalline powder or crystals or granules
Titration with HClO496.0 to 103.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only