Thermo Scientific Chemicals

p-Xylene, 99%, pure, Thermo Scientific Chemicals

Catalog number: 158770010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

p-Xylene, 99%, pure, Thermo Scientific Chemicals

Catalog number: 158770010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 158770010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
106-42-3
IUPAC Name
1,4-xylene
Molecular Formula
C8H10
InChI Key
URLKBWYHVLBVBO-UHFFFAOYSA-N
SMILES
CC1=CC=C(C)C=C1
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4940 to 1.4960 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text