p-Xylene, 99%, pure, Thermo Scientific Chemicals
p-Xylene, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Xylene, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
10 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 158770025
Price (USD)/ Each
195.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
230.00 
Save 34.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
195.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
230.00 
Save 34.35 (15%)
Add to cart
p-Xylene, 99%, pure, Thermo Scientific Chemicals
Catalog number158770025
Price (USD)/ Each
195.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
230.00 
Save 34.35 (15%)
-
Add to cart
Chemical Identifiers
CAS106-42-3
IUPAC Name1,4-xylene
Molecular FormulaC8H10
InChI KeyURLKBWYHVLBVBO-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4940 to 1.4960 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only