Thermo Scientific Chemicals

BOC-Glycine, 99+%, Thermo Scientific Chemicals

Catalog number: 158800100
10 g, Glass Bottle, Each
Thermo Scientific Chemicals

BOC-Glycine, 99+%, Thermo Scientific Chemicals

Catalog number: 158800100
10 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 158800100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4530-20-5
IUPAC Name
2-{[(tert-butoxy)carbonyl]amino}acetic acid
Molecular Formula
C7H13NO4
InChI Key
VRPJIFMKZZEXLR-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCC(O)=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
85°C to 90°C
Titration with NaOH
>=99.0 %
HPLC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text