2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 158820250
Price (USD)/ Each
37.65
Online exclusive
41.20 
Save 3.55 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.20 
Save 3.55 (9%)
Add to cart
2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
Catalog number158820250
Price (USD)/ Each
37.65
Online exclusive
41.20 
Save 3.55 (9%)
-
Add to cart
Chemical Identifiers
CAS52-51-7
IUPAC Name2-bromo-2-nitropropane-1,3-diol
Molecular FormulaC3H6BrNO4
InChI KeyLVDKZNITIUWNER-UHFFFAOYSA-N
SMILESOCC(Br)(CO)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Water=<0.5 %
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals
Titration Argentometric>=97.5 % (Ex Br)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only