Thermo Scientific Chemicals

2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: 158825000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: 158825000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 158825000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52-51-7
IUPAC Name
2-bromo-2-nitropropane-1,3-diol
Molecular Formula
C3H6BrNO4
InChI Key
LVDKZNITIUWNER-UHFFFAOYSA-N
SMILES
OCC(Br)(CO)[N+]([O-])=O
GC
>=97.5 %
Water
=<0.5 %
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder or crystals
Titration Argentometric
>=97.5 % (Ex Br)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text