Thermo Scientific Chemicals

Vinylacetic acid, tech., 90%, unstabilized

Catalog number: 158831000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Vinylacetic acid, tech., 90%, unstabilized

Catalog number: 158831000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 158831000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
625-38-7
IUPAC Name
but-3-enoic acid
Molecular Formula
C4H6O2
InChI Key
PVEOYINWKBTPIZ-UHFFFAOYSA-N
SMILES
OC(=O)CC=C
Appearance (Form)
Liquid
GC
>=88 %
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text