Thermo Scientific Chemicals

sym-Diphenylcarbazide, 98%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 158840250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

sym-Diphenylcarbazide, 98%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 158840250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 158840250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-22-7
IUPAC Name
1,3-bis(phenylamino)urea
Molecular Formula
C13H14N4O
InChI Key
KSPIHGBHKVISFI-UHFFFAOYSA-N
SMILES
O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
Appearance (Color)
White to yellow to beige to pink to
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Melting point
173°C to 176°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text