Thermo Scientific Chemicals

Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals

Catalog number: 158900010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals

Catalog number: 158900010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 158900010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2802-68-8
IUPAC Name
bis(methanol); trifluoroborane
Molecular Formula
C2H8BF3O2
InChI Key
MPRMFRXCVMCOMX-UHFFFAOYSA-N
SMILES
CO.CO.FB(F)F
Assay
50 to 53 wt% (BF3)
Appearance (Form)
Liquid
Density
(20°C) 1.210 to 1.235 kg/l
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text