Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 158902500
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.50 
Save 6.85 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.50 
Save 6.85 (15%)
Add to cart
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Catalog number158902500
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.50 
Save 6.85 (15%)
-
Add to cart
Chemical Identifiers
CAS2802-68-8
IUPAC Namebis(methanol); trifluoroborane
Molecular FormulaC2H8BF3O2
InChI KeyMPRMFRXCVMCOMX-UHFFFAOYSA-N
SMILESCO.CO.FB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay50 to 53 wt% (BF3)
Appearance (Form)Liquid
Density(20°C) 1.210 to 1.235 kg/l
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only