Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycerol, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic Drum
Catalog number 158920250
Price (USD)/ Each
878.65
Online exclusive
1034.00 
Save 155.35 (15%)
-
Add to cart
Quantity:
25 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
878.65
Online exclusive
1034.00 
Save 155.35 (15%)
Add to cart
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number158920250
Price (USD)/ Each
878.65
Online exclusive
1034.00 
Save 155.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
Molecular Weight (g/mol)92.09
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Color scale=<10 APHA
Infrared spectrumConforms
Refractive index1.4700 to 1.4750 (20°C, 589 nm)
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only