Isopropyl ether, 98+%, pure, stabilized with BHT
Isopropyl ether, 98+%, pure, stabilized with BHT
Isopropyl ether, 98+%, pure, stabilized with BHT
Isopropyl ether, 98+%, pure, stabilized with BHT
Thermo Scientific Chemicals

Isopropyl ether, 98+%, pure, stabilized with BHT

CAS: 108-20-3 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 158960010
Price (EUR)
26,78
Online Exclusive
31,50
Save 4,72 (15%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
26,78
Online Exclusive
31,50
Save 4,72 (15%)
Each
Add to cart
Chemical Identifiers
CAS108-20-3
IUPAC Name2-(propan-2-yloxy)propane
Molecular FormulaC6H14O
InChI KeyZAFNJMIOTHYJRJ-UHFFFAOYSA-N
SMILESCC(C)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Peroxides=<0.002 % (as H2O2)
Acidity=<0.005 %
Infrared spectrumConforms
Color scale=<15 APHA
GC>=98.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H336 - May cause drowsiness or dizziness

Physical Hazards

H225 - Highly flammable liquid and vapor

EU Specific Hazard Statements

EUH019 - May form explosive peroxides

EUH066 - Repeated exposure may cause skin dryness or cracking

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P312 - Call a POISON CENTER or doctor if you feel unwell

P403 + P233 - Store in a well-ventilated place. Keep container tightly closed

Share catalog number, name or link