Isopropyl ether, 98+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Isopropyl ether, 98+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropyl ether, 98+%, pure, stabilized with BHT, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number 158960100
Price (USD)/ Each
-
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-(propan-2-yloxy)propane
Molecular FormulaC6H14O
InChI KeyZAFNJMIOTHYJRJ-UHFFFAOYSA-N
SMILESCC(C)OC(C)C
Molecular Weight (g/mol)102.18
View more
SpecificationsSpecification SheetSpecification Sheet
Peroxides=<0.002 % (as H2O2)
Infrared spectrumConforms
Acidity=<0.005 %
Color scale=<15 APHA
GC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only