Thermo Scientific Chemicals

Catechol, 99+%, Thermo Scientific Chemicals

Catalog number: 158985000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Catechol, 99+%, Thermo Scientific Chemicals

Catalog number: 158985000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 158985000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-80-9
IUPAC Name
benzene-1,2-diol
Molecular Formula
C6H6O2
InChI Key
YCIMNLLNPGFGHC-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1O
Appearance (Form)
Crystalline flakes
Melting point
103°C to 106°C
Appearance
May darken on storage
Infrared spectrum
Conforms
Appearance (Color)
Off-white to grey-brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text