Cyclohexyl Bromide, 99%, Thermo Scientific Chemicals
Cyclohexyl Bromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexyl Bromide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number 159080010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebromocyclohexane
Molecular FormulaC6H11Br
InChI KeyAQNQQHJNRPDOQV-UHFFFAOYSA-N
SMILESBrC1CCCCC1
Molecular Weight (g/mol)163.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4940 to 1.4960 (20°C, 589 nm)
Water=<0.1 % (K.F.)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only