Thermo Scientific Chemicals

DL-Alanine 99%, Thermo Scientific Chemicals

Catalog number: 159091000
100 g, Plastic Bottle, Each
Thermo Scientific Chemicals

DL-Alanine 99%, Thermo Scientific Chemicals

Catalog number: 159091000
100 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 159091000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
302-72-7
IUPAC Name
2-aminopropanoic acid
Molecular Formula
C3H7NO2
InChI Key
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
SMILES
CC(N)C(O)=O
Appearance (Form)
Crystalline powder or crystals
Loss on drying
=<0.2 % (105°C, 3 hrs)
Chloride (Cl)
=<200 ppm
Iron (Fe)
=<20 ppm
Titration with HClO4
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text