Thermo Scientific Chemicals

Phenyltrimethylammonium Bromide 98%, Thermo Scientific Chemicals

Catalog number: 159161000
Each
Thermo Scientific Chemicals

Phenyltrimethylammonium Bromide 98%, Thermo Scientific Chemicals

Catalog number: 159161000
Each
Catalog number: 159161000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16056-11-4
IUPAC Name
N,N,N-trimethylanilinium bromide
Molecular Formula
C9H14BrN
InChI Key
GNMJFQWRASXXMS-UHFFFAOYSA-M
SMILES
[Br-].C[N+](C)(C)C1=CC=CC=C1
Titration Argentometric
>=97.5 %
Appearance (Form)
Crystalline powder and/or chunks
Appearance (Color)
White to blue
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator, Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text