Acetylcholine Chloride, 99%
Acetylcholine Chloride, 99%
Acetylcholine Chloride, 99%
Acetylcholine Chloride, 99%
Thermo Scientific Chemicals

Acetylcholine Chloride, 99%

CAS: 60-31-1 | C7H16ClNO2 | 181.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 159170050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS60-31-1
IUPAC Name[2-(acetyloxy)ethyl]trimethylazanium chloride
Molecular FormulaC7H16ClNO2
InChI KeyJUGOREOARAHOCO-UHFFFAOYSA-M
SMILES[Cl-].CC(=O)OCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Loss on drying=<1.0 % (105°C, 3 hrs)
Appearance (Form)Crystalline powder
Melting point146°C to 151°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link