Acetylcholine Chloride, 99%, Thermo Scientific Chemicals
Acetylcholine Chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetylcholine Chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 159171000
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
Add to cart
Acetylcholine Chloride, 99%, Thermo Scientific Chemicals
Catalog number159171000
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
CAS60-31-1
IUPAC Name[2-(acetyloxy)ethyl]trimethylazanium chloride
Molecular FormulaC7H16ClNO2
InChI KeyJUGOREOARAHOCO-UHFFFAOYSA-M
SMILES[Cl-].CC(=O)OCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Loss on drying=<1.0 % (105°C, 3 hrs)
Appearance (Form)Crystalline powder
Melting point146°C to 151°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only