Thermo Scientific Chemicals

4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 159252500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 159252500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 159252500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
74-11-3
IUPAC Name
4-chlorobenzoic acid
Molecular Formula
C7H5ClO2
InChI Key
XRHGYUZYPHTUJZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(Cl)C=C1
Appearance (Form)
Crystalline powder
HPLC
>=98.5 %
Titration with NaOH
98.5 to 101.5 %
Water
=<0.1 % (K.F.)
Melting point
240°C to 243°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text