Dimethyl oxalate, 99%, Thermo Scientific Chemicals
Dimethyl oxalate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl oxalate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 159271000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS553-90-2
IUPAC Namedimethyl oxalate
Molecular FormulaC4H6O4
InChI KeyLOMVENUNSWAXEN-UHFFFAOYSA-N
SMILESCOC(=O)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point51°C to 55°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only