Thermo Scientific Chemicals

Hydrindantin dihydrate, 96%, Thermo Scientific Chemicals

Catalog number: 159282500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Hydrindantin dihydrate, 96%, Thermo Scientific Chemicals

Catalog number: 159282500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 159282500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5950-69-6
IUPAC Name
2,2',3,3,3',3'-hexahydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1'-dione
Molecular Formula
C18H14O8
InChI Key
QHVADKNWNMILPQ-UHFFFAOYNA-N
SMILES
OC1(O)C2=CC=CC=C2C(=O)C1(O)C1(O)C(=O)C2=CC=CC=C2C1(O)O
Appearance (Color)
Light beige to pink-beige
Infrared spectrum
Conforms
HPLC
>=95 %
Loss on drying
9 % to 11 % (105°C, 3 hr)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text