Thermo Scientific Chemicals

Benzamide, 99%, Thermo Scientific Chemicals

Catalog number: 159335000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Benzamide, 99%, Thermo Scientific Chemicals

Catalog number: 159335000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 159335000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
55-21-0
IUPAC Name
benzamide
Molecular Formula
C7H7NO
InChI Key
KXDAEFPNCMNJSK-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC=CC=C1
Appearance (Color)
White to off-white
HPLC
>=98.5 %
Appearance (Form)
Crystalline powder
Melting point
125°C to 129°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text