Thermo Scientific Chemicals

4,4'-Methylenebis(cyclohexylamine), 95%, Thermo Scientific Chemicals

Catalog number: 159342500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

4,4'-Methylenebis(cyclohexylamine), 95%, Thermo Scientific Chemicals

Catalog number: 159342500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 159342500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1761-71-3
IUPAC Name
4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
Molecular Formula
C13H26N2
InChI Key
DZIHTWJGPDVSGE-UHFFFAOYSA-N
SMILES
NC1CCC(CC2CCC(N)CC2)CC1
Appearance (Form)
Liquid or low melting solid
GC
>=94 %
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text