Thermo Scientific Chemicals

Sulfur trioxide trimethylamine complex, 95%, Thermo Scientific Chemicals

Catalog number: 159351000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Sulfur trioxide trimethylamine complex, 95%, Thermo Scientific Chemicals

Catalog number: 159351000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 159351000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3162-58-1
IUPAC Name
sulfonylideneoxidane; trimethylamine
Molecular Formula
C3H9NO3S
InChI Key
DXASQZJWWGZNSF-UHFFFAOYSA-N
SMILES
CN(C)C.O=S(=O)=O
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Appearance (Form)
Powder
Titration with NaOH
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text