Sulfur trioxide trimethylamine complex, 95%, Thermo Scientific Chemicals
Sulfur trioxide trimethylamine complex, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfur trioxide trimethylamine complex, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 159355000
Price (USD)/ Each
370.00
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
370.00
Add to cart
Sulfur trioxide trimethylamine complex, 95%, Thermo Scientific Chemicals
Catalog number159355000
Price (USD)/ Each
370.00
-
Add to cart
Chemical Identifiers
CAS3162-58-1
IUPAC Namesulfonylideneoxidane; trimethylamine
Molecular FormulaC3H9NO3S
InChI KeyDXASQZJWWGZNSF-UHFFFAOYSA-N
SMILESCN(C)C.O=S(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Powder
Titration with NaOH>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only